Assay Details
Target Gene Details
Entrez Gene ID: | 237711 |
Gene Name: | echinoderm microtubule associated protein like 6 |
Gene Aliases: |
2900083P10Rik, C230094A16Rik, EMAP-6 |
Location: |
Chr.11:29742586-30026034 on Build GRCm38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eml6 | NM_146016.2 | 42 | 6778 | NP_666128.2 |
| AK081418.1 | 7 | 1129 | BAC38214.1 | |
| BC024726.1 | 14 | 2213 | AAH24726.1 |
Target Gene Details
Entrez Gene ID: | 68585 |
Gene Name: | reticulon 4 |
Gene Aliases: |
1110020G17Rik, AA407876, AA409940, AA960376, ASY, C130026I10Rik, NOGO, NSP-CL, NgA, Nogo-A, Nogo-B, Nogo-C, mKIAA0886, mKIAA4153 |
Location: |
Chr.11:29692898-29744414 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4 | NM_024226.4 | 7 | 3210 | NP_077188.1 |
| NM_194051.3 | 9 | 5497 | NP_918940.1 | |
| NM_194052.3 | 7 | 3686 | NP_918941.1 | |
| NM_194053.3 | 8 | 3743 | NP_918942.1 | |
| NM_194054.3 | 9 | 6104 | NP_918943.1 | |
| AK047253.1 | ||||
| BU698622.1 | 1 | 62 |

Genomic Map