Assay Details
Target Gene Details
Entrez Gene ID: | 192231 |
Gene Name: | hexamethylene bis-acetamide inducible 1 |
Gene Aliases: |
7330426E13Rik, CLP-1, Clp1, HIS1 |
Location: |
Chr.11:103116325-103119725 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexim1 | NM_138753.2 | 1 | 1131 | NP_620092.1 |
AK134535.1 | 1 | 1131 | BAE22173.1 | |
AY090614.1 | 1 | 1131 | AAM09026.1 | |
BC022111.1 | 1 | 651 | AAH22111.1 |
