Assay Details
Target Gene Details
Entrez Gene ID: | 15239 |
Gene Name: | HGF-regulated tyrosine kinase substrate |
Gene Aliases: |
Hgr, Hrs, ZFYVE8 |
Location: |
Chr.11:120467635-120483984 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgs | NM_001159328.1 | 22 | 2501 | NP_001152800.1 |
| NM_008244.3 | 22 | 2498 | NP_032270.3 | |
| AK046299.1 | 22 | 2459 | BAC32676.1 | |
| AK145031.1 | 22 | 2421 | BAE26195.1 | |
| AK166441.1 | 21 | 2759 | BAE38778.1 | |
| AK209070.1 | 1 | 122 | ||
| BC003239.1 | 22 | 2442 | AAH03239.1 | |
| D50050.1 | BAA08768.1 |

Genomic Map