Assay Details
Target Gene Details
Entrez Gene ID: | 268294 |
Gene Name: | zinc finger and BTB domain containing 24 |
Gene Aliases: |
BC055367, Bif1, Bsg1, ZNF450 |
Location: |
Chr.10:41450356-41465582 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb24 | NM_001277229.1 | 6 | 2356 | NP_001264158.1 |
| NM_153398.3 | 7 | 2422 | NP_700447.2 | |
| NR_102360.1 | 7 | 2417 | ||
| AK054097.1 | 7 | 2378 | BAE20695.1 | |
| AK146986.1 | 7 | 2398 | BAE27589.1 | |
| AK154467.1 | 7 | 2393 | ||
| AY210402.1 | 7 | 2396 | AAO42998.1 | |
| BC050933.1 | 7 | 2384 | AAH50933.1 | |
| BC055367.1 | 6 | 2299 | AAH55367.1 |
More Information
Set Membership: |
|

Genomic Map