Assay Details
Target Gene Details
Entrez Gene ID: | 103737 |
Gene Name: | peroxisomal biogenesis factor 12 |
Gene Aliases: |
AI451906 |
Location: |
Chr.11:83294645-83298996 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex12 | NM_134025.3 | 4 | 1237 | NP_598786.1 |
| AK035552.1 | 4 | 1238 | BAC29102.1 | |
| AK153798.1 | 3 | 1444 | BAE32186.1 | |
| AK166413.1 | 4 | 1281 | BAE38761.1 | |
| BC021800.1 | 3 | 1061 | AAH21800.1 |
More Information
Set Membership: |
|

Genomic Map