Assay Details
Target Gene Details
Entrez Gene ID: | 69806 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 11 |
Gene Aliases: |
1810074D23Rik |
Location: |
Chr.11:113244851-113650752 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a11 | NM_001166503.1 | 10 | 2203 | NP_001159975.1 |
| NM_027216.5 | 10 | 2182 | NP_081492.4 | |
| AK007986.1 | 10 | 2122 | BAB25390.1 | |
| AK036999.1 | 10 | 2129 | BAC29664.1 | |
| AK049368.1 | 10 | 2147 | BAC33713.1 | |
| AK165545.1 | 8 | 2034 | ||
| AK170673.1 | 10 | 2122 | BAE41950.1 | |
| AK208507.1 | ||||
| BC004643.1 | 7 | 1691 | AAH04643.1 | |
| BC019647.1 | 11 | 2105 | AAH19647.1 |
More Information
Set Membership: |
|

Genomic Map