Assay Details
Target Gene Details
Entrez Gene ID: | 17846 |
Gene Name: | COMM domain containing 1 |
Gene Aliases: |
AI256843, Murr1, U2/Mu |
Location: |
Chr.11:22899728-22982843 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Commd1 | NM_144514.2 | 2 | 258 | NP_653097.2 |
| AB076722.1 | 3 | 935 | BAB82376.1 | |
| AK082909.1 | 2 | 235 | ||
| AK133507.1 | 2 | 234 | BAE21693.1 | |
| BC030052.1 | 2 | 158 | AAH30052.1 | |
| BC051210.1 | 2 | 219 | AAH51210.1 | |
| BC132083.1 | 2 | 247 | ||
| BC132085.1 | 2 | 248 | ||
| BY337798.1 | 2 | 259 | ||
| D85430.1 | 2 | 228 |

Genomic Map