Assay Details
Target Gene Details
Entrez Gene ID: | 17846 |
Gene Name: | COMM domain containing 1 |
Gene Aliases: |
AI256843, Murr1, U2/Mu |
Location: |
Chr.11:22899728-22982843 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Commd1 | NM_144514.2 | 3 | 657 | NP_653097.2 |
AB076722.1 | 4 | 1334 | BAB82376.1 | |
AK082909.1 | 3 | 634 | ||
AK133507.1 | 3 | 633 | BAE21693.1 | |
BB053099.1 | 1 | 116 | ||
BC030052.1 | 3 | 557 | AAH30052.1 | |
BC051210.1 | 3 | 618 | AAH51210.1 | |
BC132083.1 | 3 | 646 | ||
BC132085.1 | 3 | 647 | ||
D85430.1 | 3 | 627 |
