Assay Details
Target Gene Details
Entrez Gene ID: | 268448 |
Gene Name: | PHD finger protein 12 |
Gene Aliases: |
2410142K10Rik, F630045O13, PF1, mKIAA1523 |
Location: |
Chr.11:77982754-78030539 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf12 | NM_174852.3 | 9 | 2048 | NP_777277.2 |
| AK050751.1 | 3 | 464 | ||
| AK089226.1 | 9 | 2111 | ||
| AK129383.1 | 6 | 1195 | BAC98193.2 | |
| AK141796.1 | 9 | 1952 | BAE24837.1 | |
| AK155354.1 | 9 | 2089 | BAE33212.1 | |
| AK170155.1 | 9 | 2136 | BAE41602.1 | |
| AK170187.1 | 8 | 1343 | BAE41622.1 | |
| BC043080.1 | 9 | 1651 | AAH43080.1 |
More Information
Set Membership: |
|

Genomic Map