Assay Details
Target Gene Details
Entrez Gene ID: | 76392 |
Gene Name: | schlafen 5, opposite strand |
Gene Aliases: |
1700003F17Rik |
Location: |
Chr.11:82942341-82960681 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slfn5os | NR_045932.1 | |||
| AK005634.1 | BAB24160.1 | |||
| BY705829.1 |
Target Gene Details
Entrez Gene ID: | 217012 |
Gene Name: | unc-45 homolog B (C. elegans) |
Gene Aliases: |
AA445617, Cmya4, D230041A13Rik, Unc45 |
Location: |
Chr.11:82911253-82943406 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc45b | NM_178680.4 | 20 | 2943 | NP_848795.3 |
| AF539793.1 | 19 | 2842 | AAO13383.1 | |
| AK052294.1 | 20 | 2870 | BAC34921.1 | |
| AK081821.1 | 20 | 2882 | BAC38342.1 | |
| AK084418.1 | 20 | 2911 | BAC39178.1 | |
| AK168968.1 | 20 | 2912 | ||
| BC084585.1 | 20 | 2876 | AAH84585.1 |
More Information
Set Membership: |
|

Genomic Map