Assay Details
Target Gene Details
Entrez Gene ID: | 22272 |
Gene Name: | ubiquinol-cytochrome c reductase, complex III subunit VII |
Gene Aliases: |
1100001F06Rik, 1500040F11Rik, 5830407L17Rik, 9.5kDa, AA959903, QP-C, Qpc, Uqcrb, c1502 |
Location: |
Chr.11:53428948-53430874 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcrq | NM_025352.2 | 1 | 147 | NP_079628.1 |
| AA510422.1 | 1 | 94 | ||
| AK002389.1 | BAB22063.1 | |||
| AK003160.1 | BAB22613.1 | |||
| AK007982.1 | 1 | 108 | BAB25388.1 | |
| AK017907.1 | 1 | 108 | ||
| BC028519.1 | 1 | 131 | AAH28519.1 | |
| CB950804.1 | 1 | 150 |
More Information
Set Membership: |
|

Genomic Map