Assay Details
Target Gene Details
Entrez Gene ID: | 54377 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 4 |
Gene Aliases: |
AI413107, AW491861 |
Location: |
Chr.11:107734780-107794464 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng4 | NM_019431.2 | 4 | 536 | NP_062304.1 |
| AJ272045.1 | 4 | 536 | CAB86386.1 | |
| AK144253.1 | 4 | 767 | BAE25799.1 | |
| BC138503.1 | 4 | 555 | ||
| BC138506.1 | 4 | 555 |
More Information
Set Membership: |
|

Genomic Map