Assay Details
Target Gene Details
Entrez Gene ID: | 15384 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A/B |
Gene Aliases: |
3010025C11Rik, CBF-A, Cgbfa, Hnrpab |
Location: |
Chr.11:51600100-51606881 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpab | NM_001048061.1 | 8 | 2318 | NP_001041526.1 |
| NM_010448.3 | 7 | 2177 | NP_034578.1 | |
| AK013384.1 | 7 | 2144 | BAB28821.1 |
Target Gene Details
Entrez Gene ID: | 72947 |
Gene Name: | 5-phosphohydroxy-L-lysine phospholyase |
Gene Aliases: |
2900006B13Rik, Agxt2l2 |
Location: |
Chr.11:51583983-51603269 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phykpl | NM_028398.2 | NP_082674.1 | ||
| AK013489.1 | ||||
| AK156866.1 | ||||
| AK160557.1 | BAE35872.1 | |||
| BC024461.1 | AAH24461.1 |
More Information
Set Membership: |
|

Genomic Map