Assay Details
Target Gene Details
Entrez Gene ID: | 75828 |
Gene Name: | HORMA domain containing 2 |
Gene Aliases: |
4930529M09Rik |
Location: |
Chr.11:4345801-4442459 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hormad2 | NM_029458.1 | 10 | 975 | NP_083734.1 |
AK015939.1 | 11 | 977 | BAB30043.1 | |
BC120781.1 | 10 | 975 | ||
BC120783.1 | 10 | 975 | ||
BC144842.1 | 10 | 975 |
