Assay Details
Target Gene Details
Entrez Gene ID: | 14230 |
Gene Name: | FK506 binding protein 10 |
Gene Aliases: |
AI325255, FKBP-10, FKBP-65, FKBP65, Fkbp-rs1, Fkbp1-rs, Fkbp6, Fkbprp |
Location: |
Chr.11:100415694-100424840 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp10 | NM_001163481.1 | NP_001156953.1 | ||
| NM_010221.2 | NP_034351.2 | |||
| AK019446.2 | ||||
| AK144185.1 | BAE25754.1 | |||
| BC029546.1 | AAH29546.1 | |||
| L07063.1 | AAC37678.1 |
Target Gene Details
Entrez Gene ID: | 68106 |
Gene Name: | 5'-nucleotidase, cytosolic IIIB |
Gene Aliases: |
2610037D24Rik, AI841843, C330027I04Rik, Nt5c3l |
Location: |
Chr.11:100422321-100441827 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5c3b | NM_026561.4 | 9 | 1321 | NP_080837.3 |
| AK082816.1 | 9 | 1307 | BAC38632.1 |
More Information
Set Membership: |
|

Genomic Map