Assay Details
Target Gene Details
Entrez Gene ID: | 217732 |
Gene Name: | CLOCK interacting protein, circadian |
Gene Aliases: |
2310044G17Rik, AI842544, AI853744, Kiaa1737, mKIAA1737 |
Location: |
Chr.12:86947041-86965364 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cipc | NM_001289429.1 | 5 | 1344 | NP_001276358.1 |
NM_001289430.1 | 4 | 1243 | NP_001276359.1 | |
NM_001289431.1 | 5 | 1417 | NP_001276360.1 | |
NM_001289432.1 | 5 | 1291 | NP_001276361.1 | |
NM_173735.3 | 4 | 1190 | NP_776096.2 | |
AK076398.1 | 1 | 264 | BAC36322.1 | |
AK122546.1 | 4 | 1119 | BAC65828.1 | |
AK148415.1 | 3 | 1120 | BAE28540.1 | |
AK154476.1 | 4 | 1131 | BAE32612.1 | |
AK160481.1 | 5 | 1344 | ||
AK160643.1 | 4 | 1116 | BAE35935.1 | |
BC026384.1 | 4 | 1122 | AAH26384.1 |
