Assay Details
Target Gene Details
Entrez Gene ID: | 78767 |
Gene Name: | EF-hand calcium binding domain 11 |
Gene Aliases: |
2610021K21Rik, 2610110A13Rik |
Location: |
Chr.12:99692752-99884445 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab11 | NM_030172.2 | 6 | 1281 | NP_084448.1 |
| AK011507.1 | 6 | 1281 | BAB27665.1 | |
| AK011836.1 | BAB27869.1 | |||
| AK035051.1 | 6 | 1261 | BAC28925.1 | |
| BC049109.1 | 6 | 1243 | AAH49109.1 |
More Information
Set Membership: |
|

Genomic Map