Assay Details
Target Gene Details
Entrez Gene ID: | 69640 |
Gene Name: | family with sequence similarity 83, member G |
Gene Aliases: |
2310040C09Rik, AI506990, wly |
Location: |
Chr.11:61684082-61709950 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83g | NM_178618.3 | 1 | 315 | NP_848733.2 |
| AK028566.1 | 1 | 262 | BAC26012.1 | |
| AK077262.1 | 2 | 351 | BAC36716.1 | |
| BC085279.1 | 1 | 276 | AAH85279.1 | |
| BY062936.1 |
Target Gene Details
Entrez Gene ID: | 109342 |
Gene Name: | solute carrier family 5 (sodium/glucose cotransporter), member 10 |
Gene Aliases: |
C330021F16Rik, SGLT5 |
Location: |
Chr.11:61672782-61720811 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a10 | NM_001033227.2 | NP_001028399.1 | ||
| AK144033.1 | BAE25665.1 | |||
| AK165337.1 | BAE38140.1 | |||
| BC141325.1 |
More Information
Set Membership: |
|

Genomic Map