Assay Details
Target Gene Details
Entrez Gene ID: | 16878 |
Gene Name: | leukemia inhibitory factor |
Gene Aliases: |
- |
Location: |
Chr.11:4257568-4272514 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lif | NM_001039537.2 | 3 | 2900 | NP_001034626.1 |
| NM_008501.2 | 4 | 2818 | NP_032527.1 | |
| AK042018.1 | 3 | 2602 | ||
| AK155959.1 | 3 | 2660 | BAE33523.1 |
More Information
Set Membership: |
|

Genomic Map