Assay Details
Target Gene Details
Entrez Gene ID: | 12409 |
Gene Name: | carbonyl reductase 2 |
Gene Aliases: |
MLCR |
Location: |
Chr.11:120729485-120732025 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbr2 | NM_007621.2 | 8 | 860 | NP_031647.1 |
| AK081816.1 | 8 | 831 | BAC38340.1 | |
| BC010758.1 | 8 | 820 | AAH10758.1 | |
| BF730701.1 | 1 | 162 | ||
| D26123.1 | 8 | 847 | BAA05120.1 | |
| X07411.1 | 8 | 817 | CAA30309.1 |
More Information
Set Membership: |
|

Genomic Map