Assay Details
Target Gene Details
Entrez Gene ID: | 245827 |
Gene Name: | FAT tumor suppressor homolog 2 (Drosophila) |
Gene Aliases: |
EMI2, Fath2, Gm523, mKIAA0811 |
Location: |
Chr.11:55250609-55336621 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fat2 | NM_001029988.2 | 20 | 11634 | NP_001025159.1 |
| AK132523.1 | 3 | 857 | BAE21217.1 | |
| AK173031.1 | 12 | 3532 | BAD32309.1 |
More Information
Set Membership: |
|

Genomic Map