Assay Details
Target Gene Details
Entrez Gene ID: | 268395 |
Gene Name: | N-methylpurine-DNA glycosylase |
Gene Aliases: |
9830006D05, AI326268, APNG, Aag, Mid1 |
Location: |
Chr.11:32226425-32232702 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpg | NM_010822.3 | 4 | 1169 | NP_034952.2 |
| AK018694.1 | 4 | 1149 | ||
| AK156067.1 | 4 | 1136 | BAE33569.1 | |
| BC014754.1 | 4 | 1163 | AAH14754.1 |
Target Gene Details
Entrez Gene ID: | 17168 |
Gene Name: | nitrogen permease regulator-like 3 |
Gene Aliases: |
Aag, CGTHBA, HS-26, HS-40, Mare, Phg, Prox1, m(alpha)RE |
Location: |
Chr.11:32231963-32267707 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nprl3 | NM_001284359.1 | 14 | 2741 | NP_001271288.1 |
| NM_001284360.1 | 12 | 2572 | NP_001271289.1 | |
| NM_181569.3 | 13 | 2702 | NP_853547.1 | |
| NR_104306.1 | 14 | 2922 | ||
| AK154884.1 | 14 | 2903 |
More Information
Set Membership: |
|

Genomic Map