Assay Details
Target Gene Details
Entrez Gene ID: | 13384 |
Gene Name: | membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3) |
Gene Aliases: |
6430514B01, Dlgh3 |
Location: |
Chr.11:101999652-102027048 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp3 | NM_007863.2 | 19 | 2131 | NP_031889.2 |
| AF079366.1 | 19 | 2082 | AAD12762.1 | |
| AK032270.1 | 18 | 3405 | ||
| BC062642.1 | 19 | 2099 | AAH62642.1 | |
| BQ444990.1 | 1 | 213 |
More Information
Set Membership: |
|

Genomic Map