Assay Details
Target Gene Details
Entrez Gene ID: | 78294 |
Gene Name: | ribosomal protein S27A |
Gene Aliases: |
0610006J14Rik, Uba52, Ubb, Ubc |
Location: |
Chr.11:29545842-29548040 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rps27a | NM_001033865.1 | 1 | 174 | NP_001029037.1 |
NM_024277.2 | NP_077239.1 | |||
AK018706.1 | BAB31357.1 |
