Assay Details
Target Gene Details
Entrez Gene ID: | 237926 |
Gene Name: | radical S-adenosyl methionine domain containing 1 |
Gene Aliases: |
B430319G23, BC056485 |
Location: |
Chr.11:94539798-94556861 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsad1 | NM_001013381.2 | 4 | 749 | NP_001013399.1 |
| AK046706.1 | 4 | 754 | ||
| AK148401.1 | 4 | 744 | ||
| BC056485.1 | 1 | 79 | AAH56485.1 | |
| BC139274.1 | 4 | 692 | ||
| BC139276.1 | 4 | 692 |
More Information
Set Membership: |
|

Genomic Map