Assay Details
Target Gene Details
Entrez Gene ID: | 66456 |
Gene Name: | RIKEN cDNA 2810001G20 gene |
Gene Aliases: |
AW045514 |
Location: |
Chr.11:64079484-64083259 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
2810001G20Rik | NR_033780.1 | 2 | 497 | |
AK005116.1 | 2 | 464 | ||
AK007927.1 | 2 | 499 | ||
AK012631.1 | 2 | 490 | ||
BU756145.1 | 1 | 421 |
