Assay Details
Target Gene Details
Entrez Gene ID: | 67684 |
Gene Name: | LUC7-like 3 (S. cerevisiae) |
Gene Aliases: |
3300001P08Rik, Luc7a |
Location: |
Chr.11:94291139-94321983 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Luc7l3 | NM_026313.1 | 8 | 1054 | NP_080589.1 |
| AK084714.1 | 3 | 441 | BAC39261.1 | |
| AK151228.1 | 8 | 1102 | BAE30220.1 | |
| AK151839.1 | 8 | 1102 | BAE30732.1 | |
| BC009092.1 | 8 | 1054 | AAH09092.1 |

Genomic Map