Assay Details
Target Gene Details
Entrez Gene ID: | 216516 |
Gene Name: | coiled-coil domain containing 157 |
Gene Aliases: |
4930562D19Rik, mKIAA1656 |
Location: |
Chr.11:4141123-4160293 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc157 | NM_001164620.1 | 12 | 2673 | NP_001158092.1 |
| NM_177616.3 | 13 | 2809 | NP_808284.2 | |
| AK143216.1 | 11 | 2329 | ||
| AK220409.1 | 13 | 2790 | BAD90260.1 | |
| BC046392.1 | 6 | 1552 | AAH46392.1 |

Genomic Map