Assay Details
Target Gene Details
Entrez Gene ID: | 16897 |
Gene Name: | lethal giant larvae homolog 1 (Drosophila) |
Gene Aliases: |
AI325176, Lgl1, Llglh, Mgl1 |
Location: |
Chr.11:60699670-60714188 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Llgl1 | NM_001159404.1 | 18 | 2802 | NP_001152876.1 |
| NM_001159405.1 | 18 | 2802 | NP_001152877.1 | |
| NM_008502.2 | 18 | 2796 | NP_032528.1 | |
| AK083672.1 | 2 | 382 | BAC38989.1 | |
| AK084386.1 | 10 | 1672 | BAC39171.1 | |
| AK159412.1 | 18 | 2668 | BAE35062.1 | |
| BC043458.1 | 4 | 707 | AAH43458.1 | |
| BC050913.1 | 18 | 2647 | AAH50913.1 | |
| BC055399.1 | 18 | 2769 | AAH55399.1 | |
| D16141.1 | 18 | 2646 | BAA03712.1 |
More Information
Set Membership: |
|

Genomic Map