Assay Details
Target Gene Details
Entrez Gene ID: | 64656 |
Gene Name: | mitochondrial ribosomal protein S23 |
Gene Aliases: |
D11Bwg1153e, MRP-S23, Rpms23, S23mt |
Location: |
Chr.11:88194106-88211507 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps23 | NM_001291270.1 | 5 | 1029 | NP_001278199.1 |
| NM_001291271.1 | 5 | 1273 | NP_001278200.1 | |
| NM_001291272.1 | 5 | 1140 | NP_001278201.1 | |
| NM_001291273.1 | 4 | 874 | NP_001278202.1 | |
| NM_024174.6 | 5 | 1045 | NP_077136.2 | |
| NR_111902.1 | 4 | 969 | ||
| AK009880.1 | 5 | 1100 | ||
| AK014375.2 | 5 | 1015 | BAB29307.2 | |
| AK168051.1 | 5 | 1239 | BAE40032.1 | |
| AK168277.1 | 5 | 1000 | BAE40223.1 | |
| AW121224.1 | 1 | 304 | ||
| BC002155.1 | 5 | 1092 | AAH02155.1 | |
| BC019980.1 | 5 | 1015 | AAH19980.1 |
More Information
Set Membership: |
|

Genomic Map