Assay Details
Target Gene Details
Entrez Gene ID: | 66787 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
4933433P14Rik |
Location: |
Chr.12:105685352-105703058 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gskip | NM_178613.3 | 2 | 186 | NP_848728.2 |
AK020785.1 | 2 | 124 | ||
AK029484.1 | 2 | 98 | BAC26470.1 | |
AK042664.1 | 2 | 147 | BAC31325.1 | |
AK079775.1 | 2 | 132 | BAC37748.1 | |
AK168962.1 | 2 | 150 | BAE40767.1 | |
AK171097.1 | 2 | 163 | BAE42246.1 | |
BC031494.1 | 2 | 154 | AAH31494.1 |
