Assay Details
Target Gene Details
Entrez Gene ID: | 66787 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
4933433P14Rik |
Location: |
Chr.12:105685352-105703058 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gskip | NM_178613.3 | 3 | 1979 | NP_848728.2 |
AK029484.1 | 4 | 1834 | BAC26470.1 | |
AK030204.1 | 1 | 1344 | BAC26840.1 | |
BC031494.1 | 3 | 1947 | AAH31494.1 |
