Assay Details
Target Gene Details
Entrez Gene ID: | 68938 |
Gene Name: | alveolar soft part sarcoma chromosome region, candidate 1 (human) |
Gene Aliases: |
1190006K01Rik, ASPC, ASPCR1, ASPL, ASPS, RCC17, TUG |
Location: |
Chr.11:120672439-120709447 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aspscr1 | NM_001164224.1 | 6 | 740 | NP_001157696.1 |
NM_026877.2 | 7 | 961 | NP_081153.1 | |
NM_198223.2 | 6 | 740 | NP_937866.1 | |
AK004509.1 | BAB23341.1 | |||
AK088197.1 | 7 | 812 | ||
AK146061.1 | 6 | 741 | BAE26870.1 | |
AK183906.1 | 1 | 170 | ||
AY349132.1 | 5 | 730 | AAR01613.1 | |
AY349133.1 | 7 | 878 | AAR01614.1 | |
BC019177.1 | 7 | 904 | AAH19177.1 | |
BC022115.1 | 7 | 908 | AAH22115.1 | |
BC031153.1 | 7 | 893 | AAH31153.1 |
