Assay Details
Target Gene Details
Entrez Gene ID: | 320534 |
Gene Name: | transmembrane protein 104 |
Gene Aliases: |
C630005D06Rik, mFLJ00021 |
Location: |
Chr.11:115187487-115247025 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem104 | NM_001033393.2 | 10 | 1454 | NP_001028565.1 |
| AK040894.1 | 10 | 1412 | ||
| AK049868.1 | 9 | 1297 | ||
| AK131112.1 | 9 | 1337 | BAD21362.1 | |
| AK155041.1 | 10 | 1431 | BAE33008.1 | |
| AK171457.1 | 10 | 1416 | BAE42466.1 | |
| AK198567.1 | ||||
| BC140976.1 | 10 | 1374 |

Genomic Map