Assay Details
Target Gene Details
Entrez Gene ID: | 268567 |
Gene Name: | transmembrane protein 229B |
Gene Aliases: |
6330442E10Rik, 9830138K24, AU019070 |
Location: |
Chr.12:78961795-79007659 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem229b | NM_178745.4 | 1 | 109 | NP_848860.1 |
| AK033403.1 | BAC28272.1 | |||
| AK053623.1 | BAC35450.1 | |||
| BC066067.1 | 1 | 109 | AAH66067.1 |

Genomic Map