Assay Details
Target Gene Details
Entrez Gene ID: | 237958 |
Gene Name: | signal peptide peptidase 2C |
Gene Aliases: |
4933407P14Rik, IMP5 |
Location: |
Chr.11:104186327-104191166 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sppl2c | NM_001082535.1 | 1 | 588 | NP_001076004.1 |
| NM_199019.2 | 1 | 588 | NP_950184.2 | |
| AK077122.1 | 1 | 589 | BAC36625.1 | |
| BC100418.1 | 1 | 561 | AAI00419.1 |
More Information
Set Membership: |
|

Genomic Map