Assay Details
Target Gene Details
Entrez Gene ID: | 237958 |
Gene Name: | signal peptide peptidase 2C |
Gene Aliases: |
4933407P14Rik, IMP5 |
Location: |
Chr.11:104186327-104191166 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sppl2c | NM_001082535.1 | 2 | 2151 | NP_001076004.1 |
| CF197856.1 | 2 | 457 |
More Information
Set Membership: |
|

Genomic Map