Assay Details
Target Gene Details
Entrez Gene ID: | 53313 |
Gene Name: | ATPase, Ca++ transporting, ubiquitous |
Gene Aliases: |
SERCA3b, Serca3 |
Location: |
Chr.11:72961169-72993043 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp2a3 | NM_001163336.1 | 1 | 147 | NP_001156808.1 |
| NM_001163337.1 | 1 | 147 | NP_001156809.1 | |
| NM_016745.3 | 1 | 147 | NP_058025.2 | |
| AK089487.1 | 1 | 147 | BAC40903.1 | |
| AK155904.1 | 1 | 148 | BAE33495.1 | |
| BC017639.1 | 1 | 61 | AAH17639.1 | |
| BC026147.1 | 1 | 93 | AAH26147.1 | |
| U49393.1 | AAB04098.1 | |||
| U49394.1 | AAB04099.1 |
More Information
Set Membership: |
|

Genomic Map