Assay Details
Target Gene Details
Entrez Gene ID: | 216797 |
Gene Name: | protease, serine 38 |
Gene Aliases: |
Gm249, Mpn2 |
Location: |
Chr.11:59372669-59376253 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss38 | NM_001045521.1 | 5 | 989 | NP_001038986.1 |
| AK145810.1 | 5 | 994 | BAE26664.1 | |
| BC147400.1 | 5 | 985 | ||
| BC147401.1 | 5 | 985 | ||
| BY757078.1 | 3 | 445 |
More Information
Set Membership: |
|

Genomic Map