Assay Details
Target Gene Details
Entrez Gene ID: | 19052 |
Gene Name: | protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform |
Gene Aliases: |
PP2A, R75353 |
Location: |
Chr.11:52098824-52122749 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2ca | NM_019411.4 | 1 | 220 | NP_062284.1 |
| AF076192.1 | 1 | 191 | AAD12587.1 | |
| AK076110.1 | 1 | 221 | BAC36190.1 | |
| AK172644.1 | 1 | 193 | BAE43111.1 | |
| AK195960.1 | ||||
| BC003856.1 | 1 | 197 | AAH03856.1 | |
| BC054458.1 | 1 | 180 | AAH54458.1 | |
| Z67745.1 | CAA91558.1 |
More Information
Set Membership: |
|

Genomic Map