Assay Details
Target Gene Details
Entrez Gene ID: | 19052 |
Gene Name: | protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform |
Gene Aliases: |
PP2A, R75353 |
Location: |
Chr.11:52098824-52122749 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2ca | NM_019411.4 | 7 | 1503 | NP_062284.1 |
| AF076192.1 | 7 | 1473 | AAD12587.1 | |
| AK076110.1 | 7 | 1504 | BAC36190.1 | |
| AK172644.1 | 7 | 1476 | BAE43111.1 | |
| BC003856.1 | 7 | 1480 | AAH03856.1 | |
| BC054458.1 | 7 | 1463 | AAH54458.1 |
More Information
Set Membership: |
|

Genomic Map