Assay Details
Target Gene Details
Entrez Gene ID: | 237979 |
Gene Name: | sidekick homolog 2 (chicken) |
Gene Aliases: |
4632412F08Rik, 5330435L01Rik, Sdk-2, mKIAA1514 |
Location: |
Chr.11:113776374-114066181 on Build GRCm38 |
Assay Gene Location: | Within Exon 45 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdk2 | NM_172800.2 | 45 | 6310 | NP_766388.2 |
AK129379.1 | 41 | 5691 | BAC98189.1 | |
AK161326.1 | 13 | 1676 | BAE36324.1 | |
AY351699.1 | 45 | 6310 | AAQ57661.1 |
