Assay Details
Target Gene Details
Entrez Gene ID: | 22412 |
Gene Name: | wingless-type MMTV integration site family, member 9B |
Gene Aliases: |
Wnt14b, Wnt15, clf, clf1, wnt-14b, wnt-15 |
Location: |
Chr.11:103727363-103749821 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wnt9b | NM_011719.4 | 3 | 477 | NP_035849.3 |
| AB073819.1 | 3 | 440 | BAB83866.1 | |
| AF469004.1 | 3 | 425 | AAL82385.1 | |
| AK052708.1 | 3 | 478 | BAC35109.1 | |
| BC110482.1 | 3 | 469 | AAI10483.1 |
More Information
Set Membership: |
|

Genomic Map