Assay Details
Target Gene Details
Entrez Gene ID: | 66601 |
Gene Name: | transmembrane and immunoglobulin domain containing 1 |
Gene Aliases: |
2010002A20Rik, Tmigd |
Location: |
Chr.11:76891246-76916587 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmigd1 | NM_025655.2 | 2 | 287 | NP_079931.1 |
AK008028.1 | 3 | 348 | BAB25419.1 | |
AK009118.1 | 2 | 287 | BAB26083.1 | |
BC022683.1 | 2 | 254 | AAH22683.1 | |
BC061082.1 | 2 | 277 | AAH61082.1 |
