Assay Details
Target Gene Details
Entrez Gene ID: | 20306 |
Gene Name: | chemokine (C-C motif) ligand 7 |
Gene Aliases: |
MCP-3, Scya7, fic, marc, mcp3 |
Location: |
Chr.11:82045712-82047525 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccl7 | NM_013654.3 | 3 | 443 | NP_038682.1 |
| AK078827.1 | 3 | 444 | BAC37413.1 | |
| BC061126.1 | 3 | 441 | AAH61126.1 | |
| BY696466.1 | 1 | 85 | ||
| L04694.1 | 3 | 405 | AAA37516.1 | |
| S50588.1 | 3 | 391 | AAA12542.1 | |
| S71251.1 | 3 | 423 | AAB30997.1 | |
| Z12297.1 | 3 | 391 | CAA78169.1 |
More Information
Set Membership: |
|

Genomic Map