Assay Details
Target Gene Details
Entrez Gene ID: | 216527 |
Gene Name: | cerebral cavernous malformation 2 |
Gene Aliases: |
BC029157 |
Location: |
Chr.11:6546887-6596761 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccm2 | NM_001190343.1 | 9 | 1233 | NP_001177272.1 |
NM_001190344.1 | 10 | 1319 | NP_001177273.1 | |
NM_146014.3 | 10 | 1407 | NP_666126.1 | |
AK155145.1 | 10 | 1381 | BAE33075.1 | |
AK166131.1 | 7 | 934 | BAE38589.1 | |
AK169727.1 | 10 | 1321 | ||
AK188818.1 | 2 | 232 | ||
AY442689.1 | 10 | 1294 | AAR29082.1 | |
BC029157.1 | 10 | 1338 | AAH29157.1 |
