Assay Details
Target Gene Details
Entrez Gene ID: | 12777 |
Gene Name: | chemokine (C-C motif) receptor 10 |
Gene Aliases: |
Cmkbr9, Gpr2 |
Location: |
Chr.11:101172922-101175695 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr10 | NM_007721.4 | 2 | 327 | NP_031747.2 |
AF208238.1 | 2 | 307 | AAF72872.1 | |
AF215982.1 | 2 | 314 | AAF63710.1 | |
AF215983.1 | 2 | 303 | AAF63711.1 | |
AK090044.1 | 2 | 330 | BAC41062.1 | |
BC117043.1 | 2 | 330 | ||
BC120542.1 | 2 | 330 |
