Assay Details
Target Gene Details
Entrez Gene ID: | 574403 |
Gene Name: | family with sequence similarity 196, member B |
Gene Aliases: |
EG574403, Gm6041 |
Location: |
Chr.11:34314822-34422642 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam196b | NM_001025382.2 | 4 | 2937 | NP_001020553.1 |
| BC072605.1 | 5 | 2598 | AAH72605.1 |

Genomic Map