Assay Details
Target Gene Details
Entrez Gene ID: | 192897 |
Gene Name: | integrin beta 4 |
Gene Aliases: |
AA407042, C230078O20, CD104 |
Location: |
Chr.11:115974725-116008412 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb4 | NM_001005608.2 | 8 | 1308 | NP_001005608.2 |
| NM_133663.2 | 8 | 1308 | NP_598424.2 | |
| AK048888.1 | 8 | 1190 | ||
| AK142648.1 | 8 | 998 | ||
| AK147424.1 | 8 | 1308 | ||
| BC059192.1 | 4 | 500 | AAH59192.1 | |
| L04678.1 | 9 | 1155 |
More Information
Set Membership: |
|

Genomic Map