Assay Details
Target Gene Details
Entrez Gene ID: | 194952 |
Gene Name: | jumonji domain containing 4 |
Gene Aliases: |
6430559I23 |
Location: |
Chr.11:59450045-59458567 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmjd4 | NM_001205068.1 | 6 | 1607 | NP_001191997.1 |
| NM_178659.6 | 6 | 1643 | NP_848774.1 | |
| NR_037992.1 | 6 | 1557 | ||
| AK032477.1 | 6 | 1531 | ||
| AK037954.1 | 6 | 1642 | BAC29907.1 | |
| AK038650.1 | 5 | 1100 | ||
| AK039116.1 | 6 | 1235 | ||
| AK047586.1 | 6 | 1558 | ||
| AK052144.1 | 6 | 1593 | BAC34856.1 | |
| AK079104.1 | 6 | 1601 | BAC37541.1 | |
| AK084669.1 | 6 | 1602 | ||
| AK145394.1 | 6 | 1621 | BAE26409.1 | |
| AK171919.1 | 6 | 1512 | ||
| BC049148.1 | 6 | 1542 | AAH49148.1 | |
| BC116707.1 | 6 | 1574 | ||
| BC118020.1 | 6 | 1487 |
More Information
Set Membership: |
|

Genomic Map